Physical hazards
PROTECTION AGAINST EVERY DAY LAB HAZARDS
Lab scientists are often exposed to a variety of chemical and physical hazards, putting them at risk of personal injury and lost productivity. Ansell’s broad range of chemical and mechanical gloves provide targeted hand protection against many of the every day hazards of a typical lab.
Downloadable Ansell
Ansell Single Use Gloves
Ansell product
Descriptions and benefits
HyFlex 11-318
Offers incredible cut protection in a thin, lightweight design for a confident grip when working with sharp objects.
ActivArmr 80-813
Offers flame and cut resistance, ideal when working with pyrophoric chemicals.
Crusader Flex 42-474
A comfortable glove with an enhanced grip, designed to handle hot objects up to 200 0C.
Chem-Tek 38-612   
Provides the best resistance to the most aggressive chemicals: aliphatic, halogenated and aromatic hydrocarbons as well as concentrated mineral acids.
Educational Assets
Chemical Exposure in Labs – Technical Paper
Smart selection of hand protection in labs – White Paper