Anti-Serotonin Rabbit Monoclonal Antibody [clone: EPR24431-201] (Alexa Fluor® 488)

Leverantör: Abcam
AB321853-10UL AB321853-100UL
ABCAAB321853-10ULEA 2650 SEK
ABCAAB321853-10UL ABCAAB321853-100UL
Anti-Serotonin Rabbit Monoclonal Antibody [clone: EPR24431-201] (Alexa Fluor® 488)
Antikroppar
Alexa Fluor® 488 Rabbit monoclonal [EPR24431-201] to Serotonin.

Observera: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Serotonin
Clonality: Monoclonal
Clone: EPR24431-201
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR