Anti-Collagen IV Rabbit Recombinant Multiclonal Antibody [clone: RM1065]

Leverantör: Abcam
AB315250-20UL AB315250-100UL AB315250-1ML
ABCAAB315250-20ULEA 2300 SEK
ABCAAB315250-20UL ABCAAB315250-100UL ABCAAB315250-1ML
Anti-Collagen IV Rabbit Recombinant Multiclonal Antibody [clone: RM1065]
Antikroppar
Rabbit recombinant multiclonal [RM1065] to Collagen IV.

Observera: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Collagen IV
Clonality: Recombinant Multiclonal
Clone: RM1065
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR