Anti-Syntenin Rabbit Monoclonal Antibody [clone: EPR28604-40]
Leverantör: Abcam
AB315342-20UL AB315342-100UL AB315342-1ML
ABCAAB315342-20ULEA 2725 SEK
ABCAAB315342-20UL ABCAAB315342-100UL ABCAAB315342-1ML
Anti-Syntenin Rabbit Monoclonal Antibody [clone: EPR28604-40]
Antikroppar
Rabbit monoclonal [EPR28604-40] to Syntenin.
Observera: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Syntenin
Clonality: Monoclonal
Clone: EPR28604-40
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse