Anti-TOX Rabbit Monoclonal Antibody [clone: EPR28108-10]
Leverantör: Abcam
AB322259-20UL AB322259-100UL AB322259-1ML
ABCAAB322259-20ULEA 2725 SEK
ABCAAB322259-20UL ABCAAB322259-100UL ABCAAB322259-1ML
Anti-TOX Rabbit Monoclonal Antibody [clone: EPR28108-10]
Antikroppar
Rabbit monoclonal [EPR28108-10] to TOX.
Observera: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: TOX
Clonality: Monoclonal
Clone: EPR28108-10
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse