Anti-CD62L Rabbit Monoclonal Antibody [clone: EPR17012-27] (Alexa Fluor® 647)

Leverantör: Abcam
AB315292-10UL AB315292-100UL
ABCAAB315292-10ULEA 2650 SEK
ABCAAB315292-10UL ABCAAB315292-100UL
Anti-CD62L Rabbit Monoclonal Antibody [clone: EPR17012-27] (Alexa Fluor® 647)
Antikroppar
Alexa Fluor® 647 Rabbit monoclonal [EPR17012-27] to CD62L.

Observera: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CD62L
Clonality: Monoclonal
Clone: EPR17012-27
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR