Anti-RAB10 (phospho T73) Mouse Monoclonal Antibody [clone: MJF-R21]

Leverantör: Abcam
AB315174-20UL AB315174-100UL AB315174-1ML
ABCAAB315174-20ULEA 1125 SEK
ABCAAB315174-20UL ABCAAB315174-100UL ABCAAB315174-1ML
Anti-RAB10 (phospho T73) Mouse Monoclonal Antibody [clone: MJF-R21]
Antikroppar
Mouse monoclonal [MJF-R21] to RAB10 (phospho T73) - Chimeric.

Observera: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: RAB10
Clonality: Monoclonal
Clone: MJF-R21
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR