Anti-VPS29 Rabbit Monoclonal Antibody [clone: EPR28122-8]
Leverantör: Abcam
AB315334-20UL AB315334-100UL AB315334-1ML
ABCAAB315334-20ULEA 2725 SEK
ABCAAB315334-20UL ABCAAB315334-100UL ABCAAB315334-1ML
Anti-VPS29 Rabbit Monoclonal Antibody [clone: EPR28122-8]
Antikroppar
Rabbit monoclonal [EPR28122-8] to VPS29.
Observera: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: VPS29
Clonality: Monoclonal
Clone: EPR28122-8
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat