Anti-IDH1+IDH2 Mouse Monoclonal Antibody [clone: MsMab-1]

Leverantör: Abcam
AB264055-100 AB264055-1MG AB264055-20UG
ABCAAB264055-100EA 7925 SEK
ABCAAB264055-100 ABCAAB264055-1MG ABCAAB264055-20UG
Anti-IDH1+IDH2 Mouse Monoclonal Antibody [clone: MsMab-1]
Antikroppar
Mouse monoclonal [MsMab-1] to IDH1 (mutated R132G) + IDH2 (mutated R172G).

Observera: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: IDH1+IDH2
Clonality: Monoclonal
Clone: MsMab-1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR