Anti-Blood Group Lewis a Mouse Monoclonal Antibody [clone: SPM 522]

Leverantör: Abcam
AB231432-1 AB231432-500 AB231432-100
ABCAAB231432-1EA 10825 SEK
ABCAAB231432-1 ABCAAB231432-500 ABCAAB231432-100
Anti-Blood Group Lewis a Mouse Monoclonal Antibody [clone: SPM 522]
Antikroppar
Mouse monoclonal [SPM 522] to Blood Group Lewis a.

Observera: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Blood Group Lewis a
Clonality: Monoclonal
Clone: SPM 522
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR