Anti-Tau (phospho T205) Rabbit Monoclonal Antibody [clone: EPR2403(2)]

Leverantör: Abcam
AB181206-100 AB181206-20UL AB181206-1ML
ABCAAB181206-100EA 7500 SEK
ABCAAB181206-100 ABCAAB181206-20UL ABCAAB181206-1ML
Anti-Tau (phospho T205) Rabbit Monoclonal Antibody [clone: EPR2403(2)]
Antikroppar
Rabbit monoclonal [EPR2403(2)] to Tau (phospho T205).

Observera: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Tau
Clonality: Monoclonal
Clone: EPR2403(2)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR