Anti-TRAP/CD40L Rabbit Monoclonal Antibody [clone: EPR20727-12]
Leverantör: Abcam
AB244895-100 AB244895-1
ABCAAB244895-100EA 10425 SEK
ABCAAB244895-100 ABCAAB244895-1
Anti-TRAP/CD40L Rabbit Monoclonal Antibody [clone: EPR20727-12]
Antikroppar
Rabbit monoclonal [EPR20727-12] to TRAP/CD40L - BSA and Azide free (Detector).
Type: Primary
Antigen: TRAP/CD40L
Clonality: Monoclonal
Clone: EPR20727-12
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human