Anti-DIMT1L Rabbit Monoclonal Antibody [clone: EPR24385-100]

Leverantör: Abcam
AB271003-100 AB271003-1ML
ABCAAB271003-100EA 7925 SEK
ABCAAB271003-100 ABCAAB271003-1ML
Anti-DIMT1L Rabbit Monoclonal Antibody [clone: EPR24385-100]
Antikroppar
Rabbit monoclonal [EPR24385-100] to DIMT1L.

Observera: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: DIMT1L
Clonality: Monoclonal
Clone: EPR24385-100
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR