Anti-CD47 Rabbit Recombinant Multiclonal Antibody [clone: RM1014]

Leverantör: Abcam
AB284138-100 AB284138-1
ABCAAB284138-100EA 11325 SEK
ABCAAB284138-100 ABCAAB284138-1
Anti-CD47 Rabbit Recombinant Multiclonal Antibody [clone: RM1014]
Antikroppar
Rabbit recombinant multiclonal [RM1014] to CD47 - BSA and Azide free.

Observera: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CD47
Clonality: Recombinant Multiclonal
Clone: RM1014
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR