Anti-IL-5 Rabbit Monoclonal Antibody [clone: EPR20012-256]

Leverantör: Abcam
AB242955-1 AB242955-100
ABCAAB242955-1EA 103375 SEK
ABCAAB242955-1 ABCAAB242955-100
Anti-IL-5 Rabbit Monoclonal Antibody [clone: EPR20012-256]
Antikroppar
Rabbit monoclonal [EPR20012-256] to IL-5 - BSA and Azide free (Capture).

Type: Primary
Antigen: IL-5
Clonality: Monoclonal
Clone: EPR20012-256
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR