Anti-MIP-1 alpha/CCL3+CCL4/MIP-1 beta+CCL3L1+CCL4L1 Rabbit Monoclonal Antibody [clone: EPR19900-275]

Leverantör: Abcam
AB242545-1 AB242545-100
ABCAAB242545-1EA 103375 SEK
ABCAAB242545-1 ABCAAB242545-100
Anti-MIP-1 alpha/CCL3+CCL4/MIP-1 beta+CCL3L1+CCL4L1 Rabbit Monoclonal Antibody [clone: EPR19900-275]
Antikroppar
Rabbit monoclonal [EPR19900-275] to MIP-1 alpha/CCL3 + CCL4/MIP-1 beta + CCL3L1 + CCL4L1 - BSA and Azide free (Capture).

Observera: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: MIP-1 alpha/CCL3+CCL4/MIP-1 beta+CCL3L1+CCL4L1
Clonality: Monoclonal
Clone: EPR19900-275
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR