Anti-Alpha Skeletal Muscle Actin Rabbit Monoclonal Antibody [clone: EPR18430]

Leverantör: Abcam
AB250775-1 AB250775-100
ABCAAB250775-1EA 101650 SEK
ABCAAB250775-1 ABCAAB250775-100
Anti-Alpha Skeletal Muscle Actin Rabbit Monoclonal Antibody [clone: EPR18430]
Antikroppar
Rabbit monoclonal [EPR18430] to Alpha Skeletal Muscle Actin - BSA and Azide free.

Observera: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Alpha Skeletal Muscle Actin
Clonality: Monoclonal
Clone: EPR18430
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR