Anti-Cytokeratin 14 Rabbit Monoclonal Antibody [clone: EPR20367-211]

Leverantör: Abcam
AB242655-100 AB242655-1
ABCAAB242655-100EA 10425 SEK
ABCAAB242655-100 ABCAAB242655-1
Anti-Cytokeratin 14 Rabbit Monoclonal Antibody [clone: EPR20367-211]
Antikroppar
Rabbit monoclonal [EPR20367-211] to Cytokeratin 14 - BSA and Azide free (Capture).

Observera: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Cytokeratin 14
Clonality: Monoclonal
Clone: EPR20367-211
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR