Anti-CD28 Rabbit Monoclonal Antibody [clone: EPR23280-63]

Leverantör: Abcam
AB246517-100 AB246517-1ML
ABCAAB246517-100EA 8475 SEK
ABCAAB246517-100 ABCAAB246517-1ML
Anti-CD28 Rabbit Monoclonal Antibody [clone: EPR23280-63]
Antikroppar
Rabbit monoclonal [EPR23280-63] to CD28.

Observera: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: CD28
Clonality: Monoclonal
Clone: EPR23280-63
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR