Anti-Calponin 1 Rabbit Monoclonal Antibody [clone: SP193]

Leverantör: Abcam
AB227667-1 AB227667-500 AB227667-100
ABCAAB227667-1EA 12775 SEK
ABCAAB227667-1 ABCAAB227667-500 ABCAAB227667-100
Anti-Calponin 1 Rabbit Monoclonal Antibody [clone: SP193]
Antikroppar
Rabbit monoclonal [SP193] to Calponin 1 - C-terminal.

Observera: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Calponin 1
Clonality: Monoclonal
Clone: SP193
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR