Anti-Surfactant protein D/SP-D Rabbit Monoclonal Antibody [clone: EPR21928-209]

Leverantör: Abcam
AB234257-100 AB234257-1
ABCAAB234257-100EA 11325 SEK
ABCAAB234257-100 ABCAAB234257-1
Anti-Surfactant protein D/SP-D Rabbit Monoclonal Antibody [clone: EPR21928-209]
Antikroppar
Rabbit monoclonal [EPR21928-209] to Surfactant protein D/SP-D - BSA and Azide free.

Observera: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Surfactant protein D/SP-D
Clonality: Monoclonal
Clone: EPR21928-209
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR