Anti-EIF2B5 Rabbit Monoclonal Antibody [clone: EPR13533]

Leverantör: Abcam
AB181033-100 AB181033-1ML
ABCAAB181033-100EA 7425 SEK
ABCAAB181033-100 ABCAAB181033-1ML
Anti-EIF2B5 Rabbit Monoclonal Antibody [clone: EPR13533]
Antikroppar
Rabbit monoclonal [EPR13533] to EIF2B5.

Observera: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: EIF2B5
Clonality: Monoclonal
Clone: EPR13533
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR