Anti-FKBP38 Rabbit Monoclonal Antibody [clone: EPR7441(2)]

Leverantör: Abcam
AB248311-100 AB248311-1
ABCAAB248311-100EA 11325 SEK
ABCAAB248311-100 ABCAAB248311-1
Anti-FKBP38 Rabbit Monoclonal Antibody [clone: EPR7441(2)]
Antikroppar
Rabbit monoclonal [EPR7441(2)] to FKBP38 - BSA and Azide free.

Observera: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: FKBP38
Clonality: Monoclonal
Clone: EPR7441(2)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR