Anti-Rho Rabbit Monoclonal Antibody [clone: Y486]

Leverantör: Abcam
AB247214-1 AB247214-100
ABCAAB247214-1EA 101650 SEK
ABCAAB247214-1 ABCAAB247214-100
Anti-Rho Rabbit Monoclonal Antibody [clone: Y486]
Antikroppar
Rabbit monoclonal [Y486] to Rho - BSA and Azide free.

Observera: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Rho
Clonality: Monoclonal
Clone: Y486
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR