Anti-VDAC2 Rabbit Polyclonal Antibody

Leverantör: Abcam
AB155803-100 AB155803-25
ABCAAB155803-100EA 7775 SEK
ABCAAB155803-100 ABCAAB155803-25
Anti-VDAC2 Rabbit Polyclonal Antibody
Antikroppar
Rabbit polyclonal to VDAC2 - C-terminal.

Observera: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: VDAC2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR