Interchangeable blocks for block heaters, digital, SB1
Leverantör: JENCONS SCIENTIFIC
357-122 357-123 357-124 357-125 357-126 357-128 357-129 357-132 357-134 357-136 357-138 357-127 357-130 357-131 357-133 357-135 357-137 357-139 357-140
JENC357-122EA 0 SEK
JENC357-122 JENC357-123 JENC357-124 JENC357-125 JENC357-126 JENC357-128 JENC357-129 JENC357-132 JENC357-134 JENC357-136 JENC357-138 JENC357-127 JENC357-130 JENC357-131 JENC357-133 JENC357-135 JENC357-137 JENC357-139 JENC357-140
Interchangeable blocks for block heaters, digital, SB1
Blocktermostater Block
Interchangeable blocks are made of high grade, non porous, anodised aluminium. Except the blocks used for plates and slides, all other blocks include positions for a block lifter and calibration thermometer.