Network vacuum pump units
Leverantör: VACUUBRAND GMBH + CO
733401 737401
181-1058EA 55675 SEK
181-1058 181-1658
Network vacuum pump units
Pumpar Membranpumpar
PC 500 LAN NT and PC 600 LAN NT. Chemistry vacuum pump units that are ready for connection, optimised for automatic, demand-sensitive vacuum generation in local vacuum networks e.g. VACUU•LAN®.
  • Chemistry diaphragm pump with high chemical resistance (parts in contact with media are made from PTFE compounds)
  • Oil-free vacuum up to ~9 mbar (PC 500 LAN) or up to ~2 mbar (PC 600 LAN)
  • High suction capacity, even at low suction pressures
  • Gas ballast standard for working with condensates
  • High solvent recovery rate
  • Easy-to-use vacuum controller VNC 1 with digital display
  • If no vacuum is required, the pump unit turns itself off automatically and turns itself back on as required
  • The switching points are freely selectable