Anti-NF-M Rabbit Polyclonal Antibody

Leverantör: ANTIBODIES.COM
A85323-100 A85323-50
ANTIA85323-100EA 5850 SEK
ANTIA85323-100 ANTIA85323-50
Anti-NF-M Rabbit Polyclonal Antibody
Antikroppar
Rabbit polyclonal antibody to NF-M for WB, ICC/IF and IHC with samples derived from Human, Horse, Bovine, Porcine, Chicken, Rat and Mouse.

  • Validated applications: WB, ICC/IF, IHC
  • Recommended dilutions: WB: 1:1000 to 1:5000, ICC/IF: 1:1000 to 1:2,500, IHC: 1:1000 to 1:2,50

Type: Primary
Antigen: NF-M
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Horse;Bovine;Porcine;Chicken;Rat;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR