Anti-IL-2 Receptor alpha Rabbit Recombinant Antibody [clone: DM148]

Leverantör: ANTIBODIES.COM
A318560-100 A318560-500
ANTIA318560-100EA 7250 SEK
ANTIA318560-100 ANTIA318560-500
Anti-IL-2 Receptor alpha Rabbit Recombinant Antibody [clone: DM148]
Antikroppar
Recombinant rabbit monoclonal [DM148] antibody to IL-2 Receptor alpha for ELISA and Flow cytometry with samples derived from human.

  • Validated applications: ELISA, Flow cytometry
  • Recommended dilutions: ELISA: 1:5000 to 10000, Flow cytometry: 1:100

Type: Primary
Antigen: IL-2 Receptor alpha
Clonality: Recombinant
Clone: DM148
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR