Anti-CD19 Mouse Monoclonal Antibody [clone: SJ25C1] (PE/Cy7)

Leverantör: AAT BIOQUEST
101911N0 101911N1 101911N2
AATB101911N0EA 3550 SEK
AATB101911N0 AATB101911N1 AATB101911N2
Anti-CD19 Mouse Monoclonal Antibody [clone: SJ25C1] (PE/Cy7)
Antikroppar
SJ25C1 is an anti-human monoclonal antibody that forms an immune complex with the CD19 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Observera: For research use only.

Type: Primary
Antigen: CD19 antigen
Clonality: Monoclonal
Clone: SJ25C1
Conjugation: PE/Cy7
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR