Anti-CD19 Mouse Monoclonal Antibody [clone: HIB19] (iFluor® 560)

Leverantör: AAT BIOQUEST
101920A0 101920A1
AATB101920A0EA 4125 SEK
AATB101920A0 AATB101920A1
Anti-CD19 Mouse Monoclonal Antibody [clone: HIB19] (iFluor® 560)
Antikroppar
HIB19 is an anti-human monoclonal antibody that recognises the CD19 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Observera: For research use only.

Type: Primary
Antigen: CD19 antigen
Clonality: Monoclonal
Clone: HIB19
Conjugation: iFluor® 560
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR