[6]-Gingerol ≥97%

Leverantör: ENZO LIFE SCIENCES AG
Fara

Synonyms: (S)-5-Hydroxy-1-(4-hydroxy-3-methoxyphenyl)decan-3-one, 1-(4ʹ-Hydroxy-3ʹ-methoxyphenyl)-5-hydroxy-3-decanone, 6-Gingerol, (S)-5-Hydroxy-1-(4-hydroxy-3-methoxyphenyl)-3-decanone, Gingerol

BMLCA4220005 BMLCA4220020
ENZOBMLCA4220005EA 1500 SEK
ENZOBMLCA4220005 ENZOBMLCA4220020
[6]-Gingerol ≥97%
[6]-Gingerol

ATPase activator.


Gingerol ([6]-gingerol), a pungent vanilloid isolated from ginger rhizomes (Zingiber officinale), is a novel and selective activator of skeletal and cardiac SR Ca²⁺-ATPase (EC₅₀ = 4 µM). It is a novel cardiotonic agent with antiplatelet activity. It induces apoptosis in HL-60 cells. Acts as an agonist at vanilloid receptors.


Soluble in 100% ethanol.

Formel: C₁₇H₂₆O₄
MW: 294,4 g/mol
Storage Temperature: Frys
MDL Number: MFCD00210507
CAS nummer: 23513-14-6

Order Now

Specifikation testresultat

Purity ≥97% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR