NBQX ≥98%

Leverantör: ENZO LIFE SCIENCES AG
Varning

Synonyms: 2,3-Dihydroxy-6-nitro-7-sulphamoylbenzo(f)-quinoxaline, 6-nitro-7-sulphamoylbenzo(f)-quinoxaline-2,3-dione, 2,3-Dihydroxy-6-nitro-7-sulphamoylbenzo[f]quinoxaline-2,3-dione, NBQX, 6-Nitro-2,3-dioxo-1,2,3,4-tetrahydrobenzo[f]quinoxaline-7-sulphonamide

ALX550272M005 ALX550272M025
ENZOALX550272M005EA 1925 SEK
ENZOALX550272M005 ENZOALX550272M025
NBQX ≥98%
NBQX

Water soluble, potent and selective AMPA/kainate receptor antagonist.


Soluble in water (5 mg/ml) or DMSO.

Formel: C₁₂H₈N₄O₆S
MW: 336,3 g/mol
Storage Temperature: Frys
MDL Number: MFCD00270054
CAS nummer: 118876-58-7

Order Now

Specifikation testresultat

Purity (HPLC) ≥98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR