Tin Protoporphyrin IX dichloride ≥95%

Leverantör: ENZO LIFE SCIENCES AG

Synonyms: TIn protoporphyrin ix dichloride

ALX430051M005 ALX430051M025
ENZOALX430051M005EA 955 SEK
ENZOALX430051M005 ENZOALX430051M025
Tin Protoporphyrin IX dichloride ≥95%
Tin Protoporphyrin IX dichloride

Potent inhibitor of heme oxygenase.


  • Heme oxygenase inhibitor


Dark red to violet solid.


Soluble in aqueous base.

Formel: C₃₄H₃₂N₄O₄Sn·2Cl
MW: 750,3 g/mol
Storage Temperature: Frys
MDL Number: MFCD00216794
CAS nummer: 14325-05-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR