Zinc protoporphyrin ≥95%

Leverantör: ENZO LIFE SCIENCES AG

Synonyms: Zinc(II) 3,3'-(3,8,13,17-tetramethyl-7,12-divinylporphyrin-2,18-diyl)dipropionate, Protoporphyrin IX zinc(II), Zinc (II) Protoporphyrin IX

ALX430049M005 ALX430049M025 ALX430049M100
ENZOALX430049M005EA 575 SEK
ENZOALX430049M005 ENZOALX430049M025 ENZOALX430049M100
Zinc protoporphyrin ≥95%
Zinc protoporphyrin

Potent and selective inhibitor of heme oxygenase (Ki=3nM), the enzyme that generates carbon monoxide (CO). Inhibits soluble guanylyl cyclase. Produces a time- and concentration-dependent inactivation of all three isoforms of nitric oxide synthase (IC50=0.8µM, 4.0µM, and 5.0µM for nNOS (NOS I), iNOS (NOS II) and eNOS (NOS III), respectively). Does not cross the blood-brain barrier. An useful tool to elucidate the role of CO in signal transduction and as a neurotransmitter.


  • Heme oxygenase inhibitor


Dark solid.


Soluble in pyridine, dimethyl formamide or DMSO. Colloidal in aqueous base;  slightly soluble in aqueous base/organic solvent mixtures.

Formel: C₃₄H₃₂N₄O₄Zn
MW: 626,034 g/mol
Storage Temperature: Frys
MDL Number: MFCD00011612
CAS nummer: 15442-64-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR