Noc-5 ≥90%

Leverantör: ENZO LIFE SCIENCES AG
Varning

Synonyms: 1-Hydroxy-2-oxo-3-(3-aminopropyl)-3-isopropyl-1-triazene

Packaged under inert gas.

ALX430017M005 ALX430017M010
ENZOALX430017M005EA 2025 SEK
ENZOALX430017M005 ENZOALX430017M010
Noc-5 ≥90%
Noc-5

Nitric oxide (NO) donor.


The half-life time of 0.1 mM NOC-5 is 25 min at 37 °C in 0.1 M PBS, pH 7.4.


Soluble in 0.1 M NaOH (10 mg/ml).


Observera: Protect from light and moisture. 

Formel: C₆H₁₆N₄O₂
Storage Temperature: Frys
CAS nummer: 146724-82-5

Order Now

Specifikation testresultat

Purity ≥90% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR