PTIO (=2-phenyl-4,4,5,5-tetramethylimidazoline-3-oxide-1-oxyl) ≥98%
Leverantör: ENZO LIFE SCIENCES AG
Varning
Synonyms: 2-Phenyl-4,4,5,5-tetramethylimidazoline-1-oxyl 3-oxide, 4,5-Dihydro-4,4,5,5-tetramethyl-2-phenyl-1H-imidazol-1-yloxy-1-oxide, 2-Phenyl-4,4,5,5-tetramethylimidazoline-3-oxide-1-oxyl, PTIO
ALX430007M025 ALX430007M100
ENZOALX430007M025EA 625 SEK
ENZOALX430007M025 ENZOALX430007M100
PTIO (=2-phenyl-4,4,5,5-tetramethylimidazoline-3-oxide-1-oxyl) ≥98%
PTIO (=2-phenyl-4,4,5,5-tetramethylimidazoline-3-oxide-1-oxyl)
Stable nitric oxide (NO) radical scavenger.
Soluble in methanol, 100% ethanol or water.
Formel: C₁₃H₁₇N₂O₂
MW: 233,29 g/mol
Storage Temperature: Frys
MDL Number: MFCD00059929
CAS nummer: 18390-00-6
Order Now
Specifikation testresultat
Purity ≥98% (Nonaqueous Titration)
Purity Detail Purified by nonaqueous titration