Teniposide ≥97%

Leverantör: ENZO LIFE SCIENCES AG

Synonyms: (5R,5aR,8aR,9S)-9-(((2R,4aR,6R,7R,8R,8aS)-7,8-Dihydroxy-2-(thiophen-2-yl)hexahydropyrano[3,2-d][1,3]dioxin-6-yl)oxy)-5-(4-hydroxy-3,5-dimethoxyphenyl)-5,5a,8a,9-tetrahydrofuro[3',4':6,7]naphtho[2,3-d][1,3]dioxol-6(8H)-one

ALX400053M025 ALX400053M100
ENZOALX400053M025EA 1100 SEK
ENZOALX400053M025 ENZOALX400053M100
Teniposide ≥97%
Teniposide

Topoisomerase II inhibitor


Glycoside-derivative of podophyllotoxin. Has cytostatic effect. Inhibits topoisomerase II. Induces apoptosis.


Semisynthetic derivative of podophyllotoxin.

Formel: C₃₂H₃₂O₁₃S
MW: 656,66 g/mol
MDL Number: MFCD00866516
CAS nummer: 29767-20-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR