Oxaliplatin ≥98%

Leverantör: ENZO LIFE SCIENCES AG
Fara

Synonyms: (trans-1,2-Cyclohexanediamine)oxalatoplatinum(II), Oxalato(trans-l-1,2-cyclohexanediamine)platinum(II)

ALX400042M005 ALX400042M025
ENZOALX400042M005EA 1750 SEK
ENZOALX400042M005 ENZOALX400042M025
Oxaliplatin ≥98%
Oxaliplatin

Antitumor agent.


Platinum analog which has wide spectrum anticancer activity. It is found to be more active against human melanoma cell lines, and has better biochemical, pharmacological and cytotoxic properties than cisplatin and carboplatin.


Slightly soluble in water (7.9 mg/ml); very slightly soluble in methanol; almost insoluble in 100% ethanol.

Formel: C₈H₁₄N₂O₄Pt
MW: 397,29 g/mol
Storage Temperature: Frys
MDL Number: MFCD00866327
CAS nummer: 61825-94-3
Merck Index: 14,06912

Order Now

Specifikation testresultat

Purity ≥98% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR