Carboplatin ≥98%
Leverantör: ENZO LIFE SCIENCES AG
|
Fara
|
Synonyms:
cis-Diammine(1,1-cyclobutanedicarboxylato)platinum(II), cis-Diamine(1,1-cyclobutanedicarboxylato)platinum(II), 1,1-Cyclobutanedicarboxylatodiammineplatinum(II)
ALX400041M025
ALX400041M100
ENZOALX400041M025EA
760
SEK
ENZOALX400041M025
ENZOALX400041M100
Carboplatin ≥98%
Carboplatin
Antitumor agent.
An analog of cisplatin.
Soluble in water.
Formel:
C₆H₁₂N₂O₄Pt MW: 371,26 g/mol Storage Temperature: Frys |
MDL Number:
MFCD00070464 CAS nummer: 41575-94-4 EINECS: 255-446-0 Merck Index: 14,01822 |
Specifikation testresultat
Purity | ≥98% (HPLC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...