Genipin (from Gardenia jasminoides ellis) ≥98%

Leverantör: ENZO LIFE SCIENCES AG

Synonyms: Methyl (1S,2R,6S)-2-Hydroxy-9-(hydroxymethyl)-3-oxabicyclo[4.3.0]nona-4,8-diene-5-carboxylate, (1R,4aS,7aS)-Methyl 1-hydroxy-7-(hydroxymethyl)-1,4a,5,7a-tetrahydrocyclopenta[c]pyran-4-carboxylate

ALX350383M025 ALX350383M100
ENZOALX350383M025EA 1150 SEK
ENZOALX350383M025 ENZOALX350383M100
Genipin (from Gardenia jasminoides ellis) ≥98%
Genipin

Cell permeable inhibitor of uncoupling protein 2 (UCP2). Increases glucose-stimulated insulin secretion, mitochondrial membrane potential and ATP levels in pancreatic island cells. Induces apoptosis. Protein cross-linking agent. Anti-inflammatory and anti-angiogenic.


  • UCP2 inhibitor


White to off-white crystalline solid.


Soluble in DMSO (50mg/ml), dimethyl formamide (25mg/ml), 100% ethanol (5mg/ml), methanol or ethyl acetate; insoluble in water.

Formel: C₁₁H₁₄O₅
MW: 226,23 g/mol
Storage Temperature: Frys
MDL Number: MFCD00888600
CAS nummer: 6902-77-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR