(-)-Epigallocatechin gallate ≥98%

Leverantör: ENZO LIFE SCIENCES AG

Synonyms: (-)-cis-3,4-Dihydro-5,7-dihydroxy-2-(3,4,5-trihydroxyphenyl)-1(2H)-benzopyran-3-yl Gallate, (2R,3R)-5,7-Dihydroxy-2-(3,4,5-trihydroxyphenyl)chroman-3-yl 3,4,5-trihydroxybenzoate, EGCG

ALX270263M010 ALX270263M050
ENZOALX270263M010EA 565 SEK
ENZOALX270263M010 ENZOALX270263M050
(-)-Epigallocatechin gallate ≥98%
(-)-Epigallocatechin gallate

Inhibitor of NOS, telomerase and Dnmt


Antitumor reagent. Antioxidant. Protects cells from lipid peroxidation and DNA damage induced by reactive free radicals. Inhibits inducible nitric oxide synthase (iNOS; NOS II). Chemopreventive anticancer agent. Induces apoptosis in human cancer cell lines. Inhibits MAP kinase mediated signalling pathways. Inhibits angiogenesis. Inhibits telomerase and DNA methyltransferase. Anti-inflammatory agent.


Isolated from green tea.

Formel: C₂₂H₁₈O₁₁
MW: 458,38 g/mol
MDL Number: MFCD00075940
CAS nummer: 989-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR