PKSI-527 ≥95%

Leverantör: ENZO LIFE SCIENCES AG

Synonyms: trans-4-Aminomethylcyclohexanecarbonyl-phenylalanyl-4-carboxymethylanilide, 2-(4-((S)-2-(trans-4-(Aminomethyl)cyclohexane-1-carboxamido)-3-phenylpropanamido)phenyl)acetic acid hydrochloride

BMLPI1470005 BMLPI1470025
ENZOBMLPI1470005EA 1225 SEK
ENZOBMLPI1470005 ENZOBMLPI1470025
PKSI-527 ≥95%
PKSI-527

Highly selective plasma kallikrein inhibitor (Ki = 0.81 µM). Can be used for affinity purification of kalikrein. Suppresses collagen-induced arthritis in a mouse model. Synergizes with NO donors producing antithrombotic effects ex vivo.


  • Kallikrein inhibitor


White solid.


Soluble in water (5 mg/ml).

Formel: C₂₅H₃₁N₃O₄·HCl
MW: 474,0 g/mol
Storage Temperature: Frys
MDL Number: MFCD01745772
CAS nummer: 128837-71-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=962f3bd8d4dfc29235977c80d9da4809873a8adf&vid=3b2d0a093e5399d5e982f156797ac49a19407174&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR