5-Hydroxydecanoate sodium salt ≥98%

Leverantör: ENZO LIFE SCIENCES AG

Synonyms: 5-Hydroxydecanoate sodium, Sodium 5-hydroxydecanoate

BMLKC1410100 BMLKC1410500
ENZOBMLKC1410100EA 965 SEK
ENZOBMLKC1410100 ENZOBMLKC1410500
5-Hydroxydecanoate sodium salt ≥98%
5-Hydroxydecanoate sodium salt

Inhibits ATP-sensitive K+ channels.


  • K+ channel blocker


White solid.


Soluble in water (25 mg/ml).

Formel: C₁₀H₁₉O₃·Na
MW: 210,3 g/mol
MDL Number: MFCD00153808
CAS nummer: 71186-53-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b17d8a3d8afe9e2c69f4d51835d131f69b96cd6e&vid=9f1f2fbdc365583fd83576daa52671a46e06c246&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR