ICRF-193 ≥95%

Leverantör: ENZO LIFE SCIENCES AG

Synonyms: Meso-4,4'-(3,2-butanediyl)-bis(2,6-piperazinedione)

BMLGR3320001 BMLGR3320005
ENZOBMLGR3320001EA 1750 SEK
ENZOBMLGR3320001 ENZOBMLGR3320005
ICRF-193 ≥95%
ICRF-193

A topoisomerase II inhibitor that does not cause DNA damage. Induces apoptosis in several cell lines including K562 and Molt-4 cells. Induces a G2 checkpoint that is associated with an ATR-dependent inhibition of polo-like kinase 1 (plk1) activity and a decrease in cyclin B1 phosphorylation.


  • Topoisomerase inhibitor


Off-white to pale yellow solid.


Soluble in DMSO (4mg/ml).

Formel: C₁₂H₁₈N₄O₄
MW: 282,3 g/mol
Smältpunkt: 298…299 °C
Storage Temperature: Frys
CAS nummer: 21416-68-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=576d1163313c1c83a8ff9598befbd64e075d322b&vid=ab7e9145d2110fa95d49ffea740a0cfa81a03783&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR