Ellipticine ≥98%
Leverantör: ENZO LIFE SCIENCES AG
Synonyms: 5,11-Dimethyl-6H-pyrido[4,3-b]carbazole
BMLGR3150010 BMLGR3150050
ENZOBMLGR3150010EA 1400 SEK
ENZOBMLGR3150010 ENZOBMLGR3150050
Ellipticine ≥98%
Ellipticine
A DNA-intercalating antitumor agent. Inhibits DNA topoisomerase II. Inhibits the phosphorylation of p53 in Lewis lung carcinoma cells and induces apoptosis. Inhibits cytochrome P-450-1A1.
  • Topoisomerase inhibitor
Bright yellow solid.
Soluble in DMSO, DMF or ethanol.
Formel: C₁₇H₁₄N₂
MW: 246,3 g/mol
Storage Temperature: Frys
MDL Number: MFCD00010524
CAS nummer: 519-23-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1952e861a593e8439f480a4cd6408388c22871e0&vid=7001ffa95da6c50564c094994ef917d10dfb1b7e&prid=web&clid=SAPDAM