Anandamide ≥98%
Leverantör: ENZO LIFE SCIENCES AG
Synonyms: N-Arachidonoylethanolamine, AEA, (all-Z)-N-(2-Hydroxyethyl)-5,8,11,14-eicosatetraenamide
BMLFA0170005 BMLFA0170025
ENZOBMLFA0170005EA 785 SEK
ENZOBMLFA0170005 ENZOBMLFA0170025
Anandamide ≥98%
Anandamide
Endogenous [1] ligand for the CB1 receptor (CB1: Ki=52nm; CB2: Ki=1930nm [2]) and TRPV1 (Ki=5.78µM [3,4]). Inhibits NF-κB activation through direct binding to IKKβ [5] and induces apoptosis independently of cannabinoid or vanilloid receptors [6]. Activates the MAP kinase (MAPK/ERK) signalling pathway [7].
  • Endogenous CB1 receptor ligand
Colorless oil.
Soluble in ethanol (e.g. at a concentration of 50 mg/ml). It is also soluble in DMSO and dimethyl formamide.
Formel: C₂₂H₃₇NO₂
MW: 347,6 g/mol
Storage Temperature: Frys, ultralågtemperatur (–80°C)
CAS nummer: 94421-68-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=71b622eced9d65ebf9c4d2a60ef9ff364c877f77&vid=3e6eabd88e05334cfcb679d2500c8a700b10d98e&prid=web&clid=SAPDAM