H-9 HCl ≥98%

Leverantör: ENZO LIFE SCIENCES AG

Synonyms: N-(2-Aminoethyl)-5-isoquinolinesulphonamide hydrochloride, N-(2-Aminoethyl)isoquinoline-5-sulphonamide hydrochloride

BMLEI1950010 BMLEI1950050
ENZOBMLEI1950010EA 1225 SEK
ENZOBMLEI1950010 ENZOBMLEI1950050
H-9 HCl ≥98%
H-9 HCl

An excellent affinity ligand for chromatographic purification of protein kinases. Inhibits protein kinases (Ki's: PKA=1.9 µM; PKC=18 µM; PKG=0.87 µM; MLCK=70 µM).


  • PKA inhibitor


White solid.


Soluble in water.

Formel: C₁₁H₁₃N₃O₂S·HCl
MW: 287,8 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD00083242
CAS nummer: 116970-50-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=209d3692b2f5e39db3cc554264e15ba1f2ec8673&vid=5144f450e435a399c768907f7544f0656c7d8813&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR