Akt inhibitor VIII ≥98%
Leverantör: ENZO LIFE SCIENCES AG
Synonyms:
Akti-1/2, 1,3-Dihydro-1-[1-[4-(6-phenyl-1H-imidazo[4,5-g]quinoxalin-7-yl)phenylmethyl]-4-piperidinyl]-2H-benzimidazol-2-one
ENZCHM1250001
ENZCHM1250005
ENZOENZCHM1250001EA
1600
SEK
ENZOENZCHM1250001
ENZOENZCHM1250005
Akt inhibitor VIII ≥98%
Akt inhibitor VIII
Potent inhibitor of Akt1, Akt2 and Akt3 activity.
A cell-permeable compound that potently, selectively and reversibly inhibits Akt1/Akt2/Akt3 activity.
Formel:
C₃₄H₂₉N₇O MW: 551,65 g/mol |
MDL Number:
MFCD09026909 CAS nummer: 612847-09-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...