KU55933 ≥99%
Leverantör: ENZO LIFE SCIENCES AG
Synonyms:
KU-55933, 2-Morpholino-6-(thianthren-1-yl)-4H-pyran-4-one
ENZCHM1500005
ENZCHM1500010
ENZOENZCHM1500005EA
955
SEK
ENZOENZCHM1500005
ENZOENZCHM1500010
KU55933 ≥99%
KU55933
Potent and selective ATP-competitive inhibitor of ATM
KU55933 is a potent and selective ATP-competitive inhibitor of ATM, with an IC50 of 13 nmol/l and a Ki of 2.2 nmol/l. KU55933 has potential for use as a new radio- and chemo-sensitizing agent for the treatment of cancer.
Formel:
C₂₁H₁₇NO₃S₂ MW: 395,49 g/mol |
MDL Number:
MFCD08276985 CAS nummer: 587871-26-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...